Product Details

SNP ID
rs113554787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3047491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGCAGAGCCCTGAGAAGTTGC[A/G]CGATGCCATCAAAGTCATGCAGAGG
Phenotype
MIM: 608482
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100128770 PubMed Links

Gene Details

Gene
LOC100128770
Gene Name
uncharacterized LOC100128770
There are no transcripts associated with this gene.

Gene
MMP25
Gene Name
matrix metallopeptidase 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022468.4 413 Missense Mutation CAC,CGC H59R NP_071913.1
XM_011522602.2 413 Intron XP_011520904.1
XM_011522604.2 413 Intron XP_011520906.1
XM_011522605.2 413 Intron XP_011520907.1
XM_017023561.1 413 Missense Mutation CAC,CGC H59R XP_016879050.1
Gene
MMP25-AS1
Gene Name
MMP25 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details