Product Details

SNP ID
rs115621754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:49469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACAGAAATGGCTTCAAACACTC[C/T]ATTTCTGACATTAGGTAAAAAGATT
Phenotype
MIM: 606007 MIM: 614403
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLR3K PubMed Links

Gene Details

Gene
POLR3K
Gene Name
RNA polymerase III subunit K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016310.4 Intron NP_057394.3
Gene
RHBDF1
Gene Name
rhomboid 5 homolog 1
There are no transcripts associated with this gene.

Gene
SNRNP25
Gene Name
small nuclear ribonucleoprotein U11/U12 subunit 25
There are no transcripts associated with this gene.

View Full Product Details