Product Details

SNP ID
rs117591819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67398106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCCGGTCCCTGCTGCCCCCAGC[A/G]GCTGTTCCAGGCTTGCCTCCCTTGC
Phenotype
MIM: 616957
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TPPP3 PubMed Links

Gene Details

Gene
TPPP3
Gene Name
tubulin polymerization promoting protein family member 3
There are no transcripts associated with this gene.

Gene
ZDHHC1
Gene Name
zinc finger DHHC-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323627.1 Intron NP_001310556.1
NM_013304.2 Intron NP_037436.1
XM_005255915.1 Intron XP_005255972.1
XM_006721185.1 Intron XP_006721248.1
XM_011523058.1 Intron XP_011521360.1
XM_011523059.1 Intron XP_011521361.1
XM_011523060.1 Intron XP_011521362.1

View Full Product Details