Product Details

SNP ID
rs113170495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67675174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCTCATAGGTTGTTCCGCTGA[A/G]GTGCCTCACACAGGTTCTGGTTGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf86 PubMed Links

Gene Details

Gene
C16orf86
Gene Name
chromosome 16 open reading frame 86
There are no transcripts associated with this gene.

Gene
ENKD1
Gene Name
enkurin domain containing 1
There are no transcripts associated with this gene.

Gene
GFOD2
Gene Name
glucose-fructose oxidoreductase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243650.1 1177 Intron NP_001230579.1
NM_030819.3 1177 Missense Mutation CCT,CTT P380L NP_110446.3
XM_006721288.3 1177 Missense Mutation CCT,CTT P275L XP_006721351.1
XM_017023738.1 1177 Missense Mutation CCT,CTT P380L XP_016879227.1

View Full Product Details