Product Details

SNP ID
rs117948983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68302546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAGTCCAAGTCAAAACCTCTAA[G/T]GTTATGCTCTGGGGGAAGTTGTTAG
Phenotype
MIM: 610087 MIM: 605641
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRMT7 PubMed Links

Gene Details

Gene
PRMT7
Gene Name
protein arginine methyltransferase 7
There are no transcripts associated with this gene.

Gene
SLC7A6
Gene Name
solute carrier family 7 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076785.2 Intron NP_001070253.1
NM_003983.5 Intron NP_003974.3
XM_011523433.1 Intron XP_011521735.1
XM_011523434.1 Intron XP_011521736.1
XM_011523438.1 Intron XP_011521740.1
XM_011523439.1 Intron XP_011521741.1
XM_017023851.1 Intron XP_016879340.1
Gene
SLC7A6OS
Gene Name
solute carrier family 7 member 6 opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032178.2 Intron NP_115554.2
XM_011523372.2 Intron XP_011521674.1

View Full Product Details