Product Details

SNP ID
rs113530943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71732980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGCCCGTGGTACAGTTGGGGGG[G/T]ACTTGCCAGCAATCACAACCTCCTT
Phenotype
MIM: 603533 MIM: 611066
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AP1G1 PubMed Links

Gene Details

Gene
AP1G1
Gene Name
adaptor related protein complex 1 gamma 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030007.1 2861 UTR 3 NP_001025178.1
NM_001128.5 2861 UTR 3 NP_001119.3
Gene
PHLPP2
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
There are no transcripts associated with this gene.

View Full Product Details