Product Details

SNP ID
rs116535181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75477686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCTTTGCCCCCAGAGCTAGGAG[A/G]TGTGGCTCACAGGCAGGGATCATCC
Phenotype
MIM: 605294
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHST6 PubMed Links

Gene Details

Gene
CHST6
Gene Name
carbohydrate sulfotransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021615.4 2938 UTR 3 NP_067628.1
XM_005255955.4 2938 UTR 3 XP_005256012.1
XM_011523085.2 2938 UTR 3 XP_011521387.1

View Full Product Details