Product Details

SNP ID
rs117371808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55515096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCTATCAGCTTTACCAACAGTTT[C/T]GTAATGGGAGTCCCAGGAAAGGAGA
Phenotype
MIM: 612040 MIM: 120360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LPCAT2 PubMed Links

Gene Details

Gene
LPCAT2
Gene Name
lysophosphatidylcholine acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017839.4 Intron NP_060309.2
XM_005256006.2 Intron XP_005256063.1
XM_011523169.2 Intron XP_011521471.1
XM_017023376.1 Intron XP_016878865.1
Gene
MMP2
Gene Name
matrix metallopeptidase 2
There are no transcripts associated with this gene.

View Full Product Details