Product Details

SNP ID
rs114157005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88577287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTGGAGGATGAGGAAAGTGCAG[C/T]CAGGGGGCCGAGCCAGGAGGAGGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZC3H18 PubMed Links

Gene Details

Gene
ZC3H18
Gene Name
zinc finger CCCH-type containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001294340.1 364 Missense Mutation GCC,GTC A55V NP_001281269.1
NM_144604.3 364 Missense Mutation GCC,GTC A55V NP_653205.3
XM_011522863.1 364 Missense Mutation GCC,GTC A55V XP_011521165.1
XM_011522865.1 364 Intron XP_011521167.1
XM_017022934.1 364 Missense Mutation GCC,GTC A55V XP_016878423.1

View Full Product Details