Product Details

SNP ID
rs114343604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:19679048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGATGCCTATACAGTAGAAAAATA[A/C]CTGCTTTATCTACTTTACAAGCAAG
Phenotype
MIM: 609523 MIM: 609833
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALDH3A2 PubMed Links

Gene Details

Gene
ALDH3A2
Gene Name
aldehyde dehydrogenase 3 family member A2
There are no transcripts associated with this gene.

Gene
SLC47A2
Gene Name
solute carrier family 47 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099646.1 Intron NP_001093116.1
NM_001256663.1 Intron NP_001243592.1
NM_152908.3 Intron NP_690872.2
XM_017024221.1 Intron XP_016879710.1
XM_017024222.1 Intron XP_016879711.1
XM_017024223.1 Intron XP_016879712.1
XM_017024224.1 Intron XP_016879713.1
XM_017024225.1 Intron XP_016879714.1
XM_017024226.1 Intron XP_016879715.1

View Full Product Details