Product Details

SNP ID
rs113089044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21001209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTTCCATGTGAATTCTTTAAGT[A/G]CAGAAAAACAACAGAAAATGAACAG
Phenotype
MIM: 612116
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105369205 PubMed Links

Gene Details

Gene
LOC105369205
Gene Name
uncharacterized LOC105369205
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011524128.2 3604 Intron XP_011522430.1
XM_011524129.2 3604 Intron XP_011522431.1
XM_011524131.2 3604 Intron XP_011522433.1
XM_017025507.1 3604 Intron XP_016880996.1
XM_017025508.1 3604 Intron XP_016880997.1
XM_017025509.1 3604 Intron XP_016880998.1
Gene
USP22
Gene Name
ubiquitin specific peptidase 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015276.1 3604 UTR 3 NP_056091.1
XM_005256575.2 3604 UTR 3 XP_005256632.1

View Full Product Details