Product Details

SNP ID
rs116479584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7310626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGCTCTTTCACATTTCTTAGTT[C/T]CTTATCCTTTCCCTTCTACCTTCCT
Phenotype
MIM: 600187 MIM: 601935 MIM: 615865
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF5A PubMed Links

Gene Details

Gene
EIF5A
Gene Name
eukaryotic translation initiation factor 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143760.1 1207 Intron NP_001137232.1
NM_001143761.1 1207 Intron NP_001137233.1
NM_001143762.1 1207 Intron NP_001137234.1
NM_001970.4 1207 Intron NP_001961.1
XM_005256509.2 1207 Intron XP_005256566.1
XM_011523710.2 1207 UTR 3 XP_011522012.1
XM_011523711.2 1207 UTR 3 XP_011522013.1
XM_011523712.2 1207 UTR 3 XP_011522014.1
XM_011523713.2 1207 UTR 3 XP_011522015.1
XM_017024300.1 1207 UTR 3 XP_016879789.1
XM_017024301.1 1207 UTR 3 XP_016879790.1
Gene
GPS2
Gene Name
G protein pathway suppressor 2
There are no transcripts associated with this gene.

Gene
NEURL4
Gene Name
neuralized E3 ubiquitin protein ligase 4
There are no transcripts associated with this gene.

View Full Product Details