Product Details

SNP ID
rs112435041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7313275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCTTTTGCAAGGACAGGGCACC[A/G]CCAGGCTGGAGGAAACCTAGGTGGG
Phenotype
MIM: 600187 MIM: 601935 MIM: 615865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF5A PubMed Links

Gene Details

Gene
EIF5A
Gene Name
eukaryotic translation initiation factor 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143760.1 817 Intron NP_001137232.1
NM_001143761.1 817 Intron NP_001137233.1
NM_001143762.1 817 Intron NP_001137234.1
NM_001970.4 817 Intron NP_001961.1
XM_005256509.2 817 Intron XP_005256566.1
XM_011523710.2 817 Intron XP_011522012.1
XM_011523711.2 817 Intron XP_011522013.1
XM_011523712.2 817 Intron XP_011522014.1
XM_011523713.2 817 Intron XP_011522015.1
XM_017024300.1 817 Intron XP_016879789.1
XM_017024301.1 817 Intron XP_016879790.1
Gene
GPS2
Gene Name
G protein pathway suppressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004489.4 817 Silent Mutation GGC,GGT G247G NP_004480.1
Gene
NEURL4
Gene Name
neuralized E3 ubiquitin protein ligase 4
There are no transcripts associated with this gene.

View Full Product Details