Product Details

SNP ID
rs115607969
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1464744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTCTCAGCGTCTTCCAACCTCT[A/G]TGGTCTAAGACGGCCTTCCCCACTC
Phenotype
MIM: 164762 MIM: 606538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRK PubMed Links

Gene Details

Gene
CRK
Gene Name
CRK proto-oncogene, adaptor protein
There are no transcripts associated with this gene.

Gene
MYO1C
Gene Name
myosin IC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080779.1 4178 UTR 3 NP_001074248.1
NM_001080950.1 4178 UTR 3 NP_001074419.1
NM_033375.4 4178 UTR 3 NP_203693.3

View Full Product Details