Product Details
- SNP ID
-
rs112169363
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:1467912 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTCAGGCTGCTGACAGAGATTCC[G/T]GAGGGGAGAGGGCAAAGGTCAGAGG
- Phenotype
-
MIM: 606538
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MYO1C
PubMed Links
Gene Details
- Gene
- MYO1C
- Gene Name
- myosin IC
View Full Product Details