Product Details

SNP ID
rs116058353
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48576930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCGCCGTGTCAGGTAGCGGTTG[C/T]AGTGAAATTCCTTCTCCAGCTCCAA
Phenotype
MIM: 142966 MIM: 142965 MIM: 610173
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HOXB3 PubMed Links

Gene Details

Gene
HOXB3
Gene Name
homeobox B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002146.4 610 Intron NP_002137.4
XM_005257277.3 610 Intron XP_005257334.1
XM_005257280.3 610 Intron XP_005257337.1
XM_005257282.3 610 Intron XP_005257339.1
XM_006721854.2 610 Intron XP_006721917.1
XM_011524707.2 610 Intron XP_011523009.1
XM_011524708.2 610 Intron XP_011523010.1
XM_011524710.1 610 Intron XP_011523012.1
XM_011524719.1 610 Intron XP_011523021.1
XM_011524720.2 610 Intron XP_011523022.1
XM_011524721.2 610 Intron XP_011523023.1
XM_011524726.2 610 Intron XP_011523028.1
XM_017024557.1 610 Intron XP_016880046.1
XM_017024558.1 610 Intron XP_016880047.1
XM_017024559.1 610 Intron XP_016880048.1
XM_017024560.1 610 Intron XP_016880049.1
XM_017024561.1 610 Intron XP_016880050.1
XM_017024562.1 610 Intron XP_016880051.1
XM_017024563.1 610 Intron XP_016880052.1
Gene
HOXB4
Gene Name
homeobox B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024015.4 610 Missense Mutation TAC,TGC Y183C NP_076920.1
Gene
MIR10A
Gene Name
microRNA 10a
There are no transcripts associated with this gene.

View Full Product Details