Product Details

SNP ID
rs118078121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63527289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCCAGGAGGACCAAGCTAGGAG[A/G]CAGCATGCAGCGGCCACCCTCTCAG
Phenotype
MIM: 608168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNH6 PubMed Links

Gene Details

Gene
KCNH6
Gene Name
potassium voltage-gated channel subfamily H member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278919.1 Intron NP_001265848.1
NM_001278920.1 Intron NP_001265849.1
NM_030779.3 Intron NP_110406.1
NM_173092.2 Intron NP_775115.1
XM_011525308.2 Intron XP_011523610.1
XM_011525309.2 Intron XP_011523611.1
XM_011525310.2 Intron XP_011523612.1
XM_011525311.1 Intron XP_011523613.1
XM_011525312.1 Intron XP_011523614.1
XM_011525313.1 Intron XP_011523615.1
XM_017025175.1 Intron XP_016880664.1
XM_017025176.1 Intron XP_016880665.1
XM_017025177.1 Intron XP_016880666.1
XM_017025178.1 Intron XP_016880667.1
XM_017025179.1 Intron XP_016880668.1
XM_017025180.1 Intron XP_016880669.1

View Full Product Details