Product Details

SNP ID
rs112218441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63828210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTCCAAGATTGAAGAACTCCAG[C/G]TGAGGACGGACTCCAGAGGGAGCTA
Phenotype
MIM: 613369 MIM: 601681 MIM: 601736
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDX42 PubMed Links

Gene Details

Gene
DDX42
Gene Name
DEAD-box helicase 42
There are no transcripts associated with this gene.

Gene
FTSJ3
Gene Name
FtsJ homolog 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017647.3 Intron NP_060117.3
Gene
PSMC5
Gene Name
proteasome 26S subunit, ATPase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199163.1 Intron NP_001186092.1
NM_002805.5 Intron NP_002796.4
Gene
SMARCD2
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2
There are no transcripts associated with this gene.

View Full Product Details