Product Details

SNP ID
rs112293979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74917133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGGGACAATCTGCAGTTTAGA[C/G]GATCCACTTTCTCCAGTGTCCACAT
Phenotype
MIM: 607827 MIM: 607696
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OTOP2 PubMed Links

Gene Details

Gene
OTOP2
Gene Name
otopetrin 2
There are no transcripts associated with this gene.

Gene
USH1G
Gene Name
USH1 protein network component sans
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282489.2 2489 UTR 3 NP_001269418.1
NM_173477.4 2489 UTR 3 NP_775748.2
XM_011524296.2 2489 UTR 3 XP_011522598.1

View Full Product Details