Product Details

SNP ID
rs111738839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74994186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCAAGGGCTCAGCCGAAACAACC[A/G]TATCTAGGAAACTTTCTGTCCGGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDR2L PubMed Links

Gene Details

Gene
CDR2L
Gene Name
cerebellar degeneration related protein 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014603.2 Intron NP_055418.2
XM_006721852.2 Intron XP_006721915.1

View Full Product Details