Product Details

SNP ID
rs115351748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75881971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTGTTTGCCAAGCCCCAGGAGC[A/G]GTTGGGAGGCCCAGGCATCAGTGCC
Phenotype
MIM: 611041
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM47 PubMed Links

Gene Details

Gene
TRIM47
Gene Name
tripartite motif containing 47
There are no transcripts associated with this gene.

Gene
TRIM65
Gene Name
tripartite motif containing 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256124.1 Intron NP_001243053.1
NM_173547.3 Intron NP_775818.2
XM_006721760.3 Intron XP_006721823.1
XM_006721761.3 Intron XP_006721824.1
XM_011524499.2 Intron XP_011522801.1
XM_011524500.2 Intron XP_011522802.1
XM_011524501.2 Intron XP_011522803.1
XM_011524502.2 Intron XP_011522804.1
XM_011524503.2 Intron XP_011522805.1
XM_017024343.1 Intron XP_016879832.1
XM_017024344.1 Intron XP_016879833.1

View Full Product Details