Product Details

SNP ID
rs114546961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76389975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAACTTGTAAACCTCTGGGCCT[A/G]TCTCTGGGCACTAGGAGAGCCCCCG
Phenotype
MIM: 603730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPHK1 PubMed Links

Gene Details

Gene
SPHK1
Gene Name
sphingosine kinase 1
There are no transcripts associated with this gene.

Gene
UBE2O
Gene Name
ubiquitin conjugating enzyme E2 O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022066.3 4821 UTR 3 NP_071349.3
XM_005257567.1 4821 UTR 3 XP_005257624.1
XM_017024934.1 4821 UTR 3 XP_016880423.1
XM_017024935.1 4821 UTR 3 XP_016880424.1

View Full Product Details