Product Details
- SNP ID
-
rs116634328
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:78170963 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGGGGTGCTGGCCTTCCTGGCCTC[A/G]GCCTTCTTCTTGGTGGTCGACGCGT
- Phenotype
-
MIM: 603926
MIM: 188300
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C17orf99
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1062668] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C17orf99
- Gene Name
- chromosome 17 open reading frame 99
There are no transcripts associated with this gene.
- Gene
- SYNGR2
- Gene Name
- synaptogyrin 2
- Gene
- TK1
- Gene Name
- thymidine kinase 1
There are no transcripts associated with this gene.
View Full Product Details