Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_170607.2 | Intron | NP_733752.1 | |||
NM_198204.1 | Intron | NP_937847.1 | |||
NM_198205.1 | Intron | NP_937848.1 | |||
XM_017024991.1 | Intron | XP_016880480.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001256014.1 | Intron | NP_001242943.1 | |||
NM_001256015.1 | Intron | NP_001242944.1 | |||
NM_001256016.1 | Intron | NP_001242945.1 | |||
NM_013290.6 | Intron | NP_037422.2 | |||
NM_016556.3 | Intron | NP_057640.1 |