Product Details

SNP ID
rs116782431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44678392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATAAAATCTGGATCCAGGTAGG[C/T]CTCTTCATTAGCTTTCTATTTCAGC
Phenotype
MIM: 616934
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC43 PubMed Links

Gene Details

Gene
CCDC43
Gene Name
coiled-coil domain containing 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099225.1 1124 UTR 3 NP_001092695.1
NM_144609.2 1124 UTR 3 NP_653210.2
Gene
MEIOC
Gene Name
meiosis specific with coiled-coil domain
There are no transcripts associated with this gene.

View Full Product Details