Product Details

SNP ID
rs113179032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44689757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTTCGCTGGGCGCCGCCATCTT[A/G]GGGTCAGGGTCCTGCAAGCCCCTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC43 PubMed Links

Gene Details

Gene
CCDC43
Gene Name
coiled-coil domain containing 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099225.1 41 UTR 5 NP_001092695.1
NM_144609.2 41 UTR 5 NP_653210.2

View Full Product Details