Product Details

SNP ID
rs117464450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44925960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGCAAAGGTGTGTTGGCACTAA[C/T]TGCTCCCAGGTAAGGAAGGCAGGTC
Phenotype
MIM: 614570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF18B PubMed Links

Gene Details

Gene
KIF18B
Gene Name
kinesin family member 18B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001264573.1 2848 UTR 3 NP_001251503.1
NM_001265577.1 2848 UTR 3 NP_001252506.1
XM_011524385.2 2848 UTR 3 XP_011522687.1
XM_011524386.2 2848 UTR 3 XP_011522688.1
XM_011524387.2 2848 UTR 3 XP_011522689.1
XM_011524388.2 2848 UTR 3 XP_011522690.1
XM_011524389.2 2848 UTR 3 XP_011522691.1
XM_011524390.2 2848 UTR 3 XP_011522692.1
XM_011524391.2 2848 Intron XP_011522693.1
Gene
MIR6783
Gene Name
microRNA 6783
There are no transcripts associated with this gene.

View Full Product Details