Product Details

SNP ID
rs112913523
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28360548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAGGATACAAGACATGGTGGGA[A/C]TCTCAGCGACCTGGGAAAGCAAGGT
Phenotype
MIM: 611519 MIM: 610975 MIM: 616815 MIM: 193190
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
MIR4723 PubMed Links

Gene Details

Gene
MIR4723
Gene Name
microRNA 4723
There are no transcripts associated with this gene.

Gene
POLDIP2
Gene Name
DNA polymerase delta interacting protein 2
There are no transcripts associated with this gene.

Gene
SEBOX
Gene Name
SEBOX homeobox
There are no transcripts associated with this gene.

Gene
TMEM199
Gene Name
transmembrane protein 199
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152464.2 485 Missense Mutation AAT,ACT N132T NP_689677.1
Gene
VTN
Gene Name
vitronectin
There are no transcripts associated with this gene.

View Full Product Details