Product Details

SNP ID
rs115403405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:31504925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTTTGTCCCCATAAAGACTCCT[A/G]CTTCTACCTGTCTATCTCATTGCCA
Phenotype
MIM: 125671
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSG2 PubMed Links

Gene Details

Gene
DSG2
Gene Name
desmoglein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001943.4 Intron NP_001934.2

View Full Product Details