Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004775.3 | 1928 | UTR 3 | NP_004766.2 | ||
XM_005258387.4 | 1928 | UTR 3 | XP_005258444.1 | ||
XM_006722578.3 | 1928 | UTR 3 | XP_006722641.1 | ||
XM_006722579.3 | 1928 | UTR 3 | XP_006722642.1 | ||
XM_017026088.1 | 1928 | UTR 3 | XP_016881577.1 | ||
XM_017026089.1 | 1928 | UTR 3 | XP_016881578.1 | ||
XM_017026090.1 | 1928 | UTR 3 | XP_016881579.1 | ||
XM_017026091.1 | 1928 | UTR 3 | XP_016881580.1 | ||
XM_017026092.1 | 1928 | UTR 3 | XP_016881581.1 |