Product Details

SNP ID
rs116403100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:32092785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCCATCCCCCGCCATGGCCGA[C/G]GACCTCTCTGCGGCCACGTCCTACA
Phenotype
MIM: 610432 MIM: 616319
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNF125 PubMed Links

Gene Details

Gene
RNF125
Gene Name
ring finger protein 125
There are no transcripts associated with this gene.

Gene
RNF138
Gene Name
ring finger protein 138
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191324.1 467 Missense Mutation GAC,GAG D3E NP_001178253.2
NM_016271.4 467 Missense Mutation GAC,GAG D3E NP_057355.2
NM_198128.2 467 Missense Mutation GAC,GAG D3E NP_937761.1
XM_005258285.1 467 Intron XP_005258342.1
XM_005258286.1 467 Missense Mutation GAC,GAG D3E XP_005258343.1

View Full Product Details