Product Details

SNP ID
rs114328900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35991626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCAAGTCCCAAAAGTTGACTA[A/C]AAACTTTGGAATAAGCCATGTATTA
Phenotype
MIM: 610347
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPRD1A PubMed Links

Gene Details

Gene
RPRD1A
Gene Name
regulation of nuclear pre-mRNA domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303411.1 2654 UTR 3 NP_001290340.1
NM_001303412.1 2654 Intron NP_001290341.1
NM_001303413.1 2654 Intron NP_001290342.1
NM_018170.4 2654 UTR 3 NP_060640.2
XM_011526048.1 2654 Intron XP_011524350.1
XM_011526049.1 2654 UTR 3 XP_011524351.1
XM_011526050.1 2654 UTR 3 XP_011524352.1
XM_017025808.1 2654 Intron XP_016881297.1

View Full Product Details