Product Details

SNP ID
rs115862835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:47108576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAACAAAAGAGGGTTAAAAAGC[G/T]ATCAGCACTGACTGGTGTCTACCGA
Phenotype
MIM: 614697
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HDHD2 PubMed Links

Gene Details

Gene
HDHD2
Gene Name
haloacid dehalogenase like hydrolase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318765.1 1095 UTR 3 NP_001305694.1
NM_032124.4 1095 UTR 3 NP_115500.1
XM_005258371.4 1095 Intron XP_005258428.1
XM_006722557.2 1095 UTR 3 XP_006722620.1
XM_011526227.2 1095 UTR 3 XP_011524529.1
XM_011526228.2 1095 UTR 3 XP_011524530.1
XM_017026039.1 1095 UTR 3 XP_016881528.1
XM_017026040.1 1095 Intron XP_016881529.1
Gene
KATNAL2
Gene Name
katanin catalytic subunit A1 like 2
There are no transcripts associated with this gene.

View Full Product Details