Product Details

SNP ID
rs112636024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37634987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGAGTTCTAATAATTATTAAGT[C/T]ACATTATGAATTCGCTGATGGTAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP30 PubMed Links

Gene Details

Gene
ZFP30
Gene Name
ZFP30 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320666.1 1876 Silent Mutation NP_001307595.1
NM_001320667.1 1876 Silent Mutation NP_001307596.1
NM_001320668.1 1876 Silent Mutation NP_001307597.1
NM_001320669.1 1876 Silent Mutation NP_001307598.1
NM_001320670.1 1876 Silent Mutation NP_001307599.1
NM_001320671.1 1876 Silent Mutation NP_001307600.1
NM_014898.2 1876 Silent Mutation NP_055713.1

View Full Product Details