Product Details

SNP ID
rs114055559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46688346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAGTCAGGTCCAGGGAGCCTGA[C/T]TGATCATTTATTTTTTATAATAAAA
Phenotype
MIM: 607074
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRKD2 PubMed Links

Gene Details

Gene
PRKD2
Gene Name
protein kinase D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079880.1 Intron NP_001073349.1
NM_001079881.1 Intron NP_001073350.1
NM_001079882.1 Intron NP_001073351.1
NM_016457.4 Intron NP_057541.2
XM_005258716.2 Intron XP_005258773.2

View Full Product Details