Product Details

SNP ID
rs116562057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50874553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGTCCAGCCACCAACCCGCTAA[C/T]GCAGGGAATAGCTACAGAATTGCCA
Phenotype
MIM: 147960
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLK2 PubMed Links

Gene Details

Gene
KLK2
Gene Name
kallikrein related peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002231.2 Intron NP_001002231.1
NM_001256080.1 Intron NP_001243009.1
NM_005551.4 Intron NP_005542.1
Gene
KLKP1
Gene Name
kallikrein pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details