Product Details

SNP ID
rs113145387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51713684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACGTAGTTAGCGGCCAGCTTCC[A/G]CCGGCCCGAGGTGCCCCAGCCACTC
Phenotype
MIM: 601463
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAS1 PubMed Links

Gene Details

Gene
HAS1
Gene Name
hyaluronan synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297436.1 1390 Missense Mutation CGG,TGG R493W NP_001284365.1
NM_001523.3 1390 Missense Mutation CGG,TGG R494W NP_001514.2
XM_011526884.2 1390 UTR 3 XP_011525186.1
Gene
SPACA6
Gene Name
sperm acrosome associated 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316972.1 1390 Intron NP_001303901.1
NM_001316994.1 1390 Intron NP_001303923.1

View Full Product Details