Product Details

SNP ID
rs115528482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52582682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTACTCACACAAAAACGGGAAG[A/T]ACACACAAGAGAAAAATCTTTCCAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF701 PubMed Links

Gene Details

Gene
ZNF701
Gene Name
zinc finger protein 701
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172655.1 752 Missense Mutation GAA,GTA E274V NP_001166126.1
NM_018260.2 752 Missense Mutation GAA,GTA E208V NP_060730.2
XM_011527092.1 752 Intron XP_011525394.1

View Full Product Details