Product Details
- SNP ID
-
rs111838811
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:54452156 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATGGCATGGAGACGCCGATGCA[C/T]GAGAACCCGGAGTGGGAGAAGGCCC
- Phenotype
-
MIM: 616575
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LENG8
PubMed Links
Gene Details
- Gene
- LENG8
- Gene Name
- leukocyte receptor cluster member 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052925.3 |
306 |
Silent Mutation |
CAC,CAT |
H34H |
NP_443157.1 |
XM_005278248.4 |
306 |
Silent Mutation |
CAC,CAT |
H34H |
XP_005278305.1 |
XM_005278249.4 |
306 |
Silent Mutation |
CAC,CAT |
H34H |
XP_005278306.1 |
XM_005278250.4 |
306 |
Silent Mutation |
CAC,CAT |
H16H |
XP_005278307.1 |
XM_005278252.4 |
306 |
Silent Mutation |
CAC,CAT |
H34H |
XP_005278309.1 |
XM_006722996.3 |
306 |
Silent Mutation |
CAC,CAT |
H16H |
XP_006723059.1 |
XM_006722997.3 |
306 |
Silent Mutation |
CAC,CAT |
H16H |
XP_006723060.1 |
XM_011526413.2 |
306 |
Silent Mutation |
CAC,CAT |
H34H |
XP_011524715.1 |
XM_011526414.2 |
306 |
Silent Mutation |
CAC,CAT |
H34H |
XP_011524716.1 |
XM_011526415.2 |
306 |
Silent Mutation |
CAC,CAT |
H34H |
XP_011524717.1 |
- Gene
- LENG8-AS1
- Gene Name
- LENG8 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- LENG9
- Gene Name
- leukocyte receptor cluster member 9
There are no transcripts associated with this gene.
View Full Product Details