Product Details

SNP ID
rs116124223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56177661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGCCACGGCACCCAGTCTGGGA[C/T]GACACTTTGAAAACCTTTGATCACA
Phenotype
MIM: 611178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALP PubMed Links

Gene Details

Gene
GALP
Gene Name
galanin like peptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145546.1 Intron NP_001139018.1
NM_033106.3 Intron NP_149097.1

View Full Product Details