Product Details

SNP ID
rs117485692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57368291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAGCAACGAGTGTTTGACACCG[A/G]TAAGAGTCCTATGAGGTGGGAGCCA
Phenotype
MIM: 613840
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC2B PubMed Links

Gene Details

Gene
TRAPPC2B
Gene Name
trafficking protein particle complex 2B
There are no transcripts associated with this gene.

Gene
ZNF304
Gene Name
zinc finger protein 304
There are no transcripts associated with this gene.

Gene
ZNF547
Gene Name
zinc finger protein 547
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173631.3 Intron NP_775902.2

View Full Product Details