Product Details

SNP ID
rs115645345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57633923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCACAGGTTAGACAGTCGACCCA[A/C]AGTTACGTGGCTCTGGGCCGGGGCA
Phenotype
MIM: 604076 MIM: 601856
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF134 PubMed Links

Gene Details

Gene
ZNF134
Gene Name
zinc finger protein 134
There are no transcripts associated with this gene.

Gene
ZNF211
Gene Name
zinc finger protein 211
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265597.2 Intron NP_001252526.1
NM_001265598.2 Intron NP_001252527.1
NM_001265599.2 Intron NP_001252528.1
NM_001265600.2 Intron NP_001252529.1
NM_001322306.1 Intron NP_001309235.1
NM_006385.4 Intron NP_006376.2
NM_198855.3 Intron NP_942152.1

View Full Product Details