Product Details

SNP ID
rs112124650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35536869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGAAGTAGAGCCCCAGCCACAA[C/T]CAGAGCCCACACCAGTCAGGGAGGA
Phenotype
MIM: 609169 MIM: 609969 MIM: 613585
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAPDHS PubMed Links

Gene Details

Gene
GAPDHS
Gene Name
glyceraldehyde-3-phosphate dehydrogenase, spermatogenic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014364.4 240 Missense Mutation CCA,TCA P42S NP_055179.1
XM_017026579.1 240 Intron XP_016882068.1
Gene
SBSN
Gene Name
suprabasin
There are no transcripts associated with this gene.

Gene
TMEM147
Gene Name
transmembrane protein 147
There are no transcripts associated with this gene.

Gene
TMEM147-AS1
Gene Name
TMEM147 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details