Product Details

SNP ID
rs112057489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2978245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTGCCTTGAAGATGACCTCTAG[A/G]GACCAGCCCAGACCCAAGGGCCCCC
Phenotype
MIM: 612399
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TLE6 PubMed Links

Gene Details

Gene
TLE6
Gene Name
transducin like enhancer of split 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143986.1 130 Silent Mutation AGA,AGG R4R NP_001137458.1
NM_024760.2 130 Intron NP_079036.1
XM_005259645.2 130 Silent Mutation AGA,AGG R4R XP_005259702.1
XM_011528300.2 130 Silent Mutation AGA,AGG R4R XP_011526602.1
XM_011528301.1 130 Intron XP_011526603.1
XM_011528306.1 130 Intron XP_011526608.1

View Full Product Details