Product Details

SNP ID
rs116973923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3480072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGAGCGGTTCACAAAAGAGGC[A/G]GGGACTCAGATGGGGAGGGGCTTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMIM24 PubMed Links

Gene Details

Gene
SMIM24
Gene Name
small integral membrane protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136503.1 Intron NP_001129975.1

View Full Product Details