Product Details

SNP ID
rs111334655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3573794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGGGCGAGAAGGGGTCCCACGA[A/G]GAGGAGGTGAGAGTCCCTGCGCTGA
Phenotype
MIM: 605535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMG20B PubMed Links

Gene Details

Gene
HMG20B
Gene Name
high mobility group 20B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006339.2 209 Silent Mutation GAA,GAG E47E NP_006330.2
XM_017026144.1 209 Silent Mutation GAA,GAG E47E XP_016881633.1

View Full Product Details