Product Details

SNP ID
rs116240312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:678410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTGCCTCGGGACCCTGTTTTG[A/G]GAGGATGCCTCCCACATCAGACTAG
Phenotype
MIM: 605343
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FSTL3 PubMed Links

Gene Details

Gene
FSTL3
Gene Name
follistatin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005860.2 Intron NP_005851.1
Gene
PRSS57
Gene Name
protease, serine 57
There are no transcripts associated with this gene.

View Full Product Details