Product Details

SNP ID
rs117422046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6466330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGAAGCCCCACTGTGGGGATC[A/G]GATCACCAAAAGTGGCAGATGTGTG
Phenotype
MIM: 609737 MIM: 613634 MIM: 608746
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRB3 PubMed Links

Gene Details

Gene
CRB3
Gene Name
crumbs 3, cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139161.4 Intron NP_631900.1
NM_174881.3 Intron NP_777377.1
NM_174882.2 Intron NP_777378.1
XM_017027471.1 Intron XP_016882960.1
Gene
DENND1C
Gene Name
DENN domain containing 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290331.1 Intron NP_001277260.1
NM_024898.3 Intron NP_079174.2
XM_006722905.2 Intron XP_006722968.2
XM_006722906.3 Intron XP_006722969.1
XM_011528318.2 Intron XP_011526620.1
Gene
SLC25A23
Gene Name
solute carrier family 25 member 23
There are no transcripts associated with this gene.

View Full Product Details