Product Details

SNP ID
rs121434423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18868916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCACGTACAGCCGCCGCGCGCGA[T/C]AAGCGCCCCCGGGGCCGCCGCCCAA
Phenotype
MIM: 606919 MIM: 602880 MIM: 601430
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CERS1 PubMed Links

Gene Details

Gene
CERS1
Gene Name
ceramide synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290265.1 2194 Intron NP_001277194.1
NM_021267.4 2194 UTR 3 NP_067090.1
NM_198207.2 2194 Intron NP_937850.1
Gene
GDF1
Gene Name
growth differentiation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001492.5 2194 Missense Mutation TAT,TGT Y267C NP_001483.3
Gene
UPF1
Gene Name
UPF1, RNA helicase and ATPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297549.1 2194 Intron NP_001284478.1
NM_002911.3 2194 Intron NP_002902.2
XM_017027105.1 2194 Intron XP_016882594.1
XM_017027106.1 2194 Intron XP_016882595.1

View Full Product Details