Product Details

SNP ID
rs115128157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11349102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGGGTGGGGCTCAGGGCCTTC[A/G]CCACATGAGGCTGCCCCCTCCCCCA
Phenotype
MIM: 604350 MIM: 613771
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC159 PubMed Links

Gene Details

Gene
CCDC159
Gene Name
coiled-coil domain containing 159
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080503.2 Intron NP_001073972.2
XM_006722643.1 Intron XP_006722706.1
XM_011527678.2 Intron XP_011525980.2
XM_017026255.1 Intron XP_016881744.1
XM_017026256.1 Intron XP_016881745.1
XM_017026257.1 Intron XP_016881746.1
XM_017026258.1 Intron XP_016881747.1
XM_017026259.1 Intron XP_016881748.1
XM_017026260.1 Intron XP_016881749.1
XM_017026261.1 Intron XP_016881750.1
XM_017026262.1 Intron XP_016881751.1
XM_017026263.1 Intron XP_016881752.1
XM_017026264.1 Intron XP_016881753.1
Gene
PLPPR2
Gene Name
phospholipid phosphatase related 2
There are no transcripts associated with this gene.

Gene
RAB3D
Gene Name
RAB3D, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
TMEM205
Gene Name
transmembrane protein 205
There are no transcripts associated with this gene.

View Full Product Details